Glycine, Ultrapure

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

BIG0503-1KG BIG0503-500G BIG0503-100G
APOSBIG0503-1KGEA 116 GBP
APOSBIG0503-1KG APOSBIG0503-500G APOSBIG0503-100G
Glycine, Ultrapure
Glycine

  • Plant tissue culture tested

Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 240…245 °C (dec.)
Density: 1.595 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR