D-(-)-Lyxose

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

BIFS0109-1G BIFS0109-5G BIFS0109-10G
APOSBIFS0109-1GEA 21 GBP
APOSBIFS0109-1G APOSBIFS0109-5G APOSBIFS0109-10G
D-(-)-Lyxose
D(-)-Lyxose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 108…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00064362
CAS Number: 1114-34-7
EINECS: 214-212-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR