Vatalanib dihydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N-(4-Chlorophenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine dihydrochloride, Vatalanib 2HCl

BIFK0044-25MG BIFK0044-10MG
APOSBIFK0044-25MGEA 59.9 GBP
APOSBIFK0044-25MG APOSBIFK0044-10MG
Vatalanib dihydrochloride
Vatalanib dihydrochloride
Formula: C₂₀H₁₇Cl₃N₄
Melting Pt: 284…286 °C
Storage Temperature: Freezer
MDL Number: MFCD08458964
CAS Number: 212141-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR