Methyl-β-cyclodextrin (mixture of several Methylated) (DS~12)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methylated-β-Cyclodextrin, Methyl-β-cyclodextrin

BICY1002-25G BICY1002-5G BICY1002-100G
APOSBICY1002-25GEA 195 GBP
APOSBICY1002-25G APOSBICY1002-5G APOSBICY1002-100G
Methyl-β-cyclodextrin (mixture of several Methylated) (DS~12)
Methyl-beta-cyclodextrin (mixture of several Methylated)
MW: 1303.3000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074980
CAS Number: 128446-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR