(S)-Boc-α-methyl-phenylglycine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N,(S)-Boc-α-methyl-phenylglycine

BICR126-1G BICR126-5G
APOSBICR126-1GEA 321 GBP
APOSBICR126-1G APOSBICR126-5G
(S)-Boc-α-methyl-phenylglycine
(S)-Boc-α-methyl-phenylglycine
Formula: C₁₄H₁₉NO₄
Storage Temperature: Freezer
MDL Number: MFCD11052916
CAS Number: 802541-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR