4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BICL4353-1G BICL4353-5G BICL4353-2G
APOSBICL4353-1GEA 263 GBP
APOSBICL4353-1G APOSBICL4353-5G APOSBICL4353-2G
4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside
Formula: C₂₄H₂₆O₇S
Storage Temperature: Ambient
MDL Number: MFCD28397803
CAS Number: 219518-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR