DST (Disuccinimidyl tartrate)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

BICL108-250MG
APOSBICL108-250MGEA 268 GBP
APOSBICL108-250MG
DST (Disuccinimidyl tartrate)
DST (Disuccinimidyl tartrate)

Primary amine reactive -Periodate cleavable


Ref: Farries,T.C.and Atkinson, J.P. (1989) J.Immun.142, 842-847Ref: Park,L.S,et al. (1986) J.Biol.Chem.261, 205-210

Formula: C₁₂H₁₂N₂O₁₀
Storage Temperature: Freezer
MDL Number: MFCD03788224
CAS Number: 77658-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR