Fmoc-δ-azido-Nva-OH ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Fmoc-Orn(N3)

BIBP1083-250MG BIBP1083-1G BIBP1083-5G
APOSBIBP1083-250MGEA 94 GBP
APOSBIBP1083-250MG APOSBIBP1083-1G APOSBIBP1083-5G
Fmoc-δ-azido-Nva-OH ≥98%
Fmoc-δ-azido-Nva-OH
Formula: C₂₀H₂₀N₄O₄
Storage Temperature: Refrigerator
CAS Number: 1097192-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR