H-L-Dap(Boc)-OMe·HCl

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: H-Dapa(Boc)-OMe.HCl, H-Dap(Boc)-OMe·HCl

BIBA1021-1G BIBA1021-5G BIBA1021-10G
APOSBIBA1021-1GEA 73.5 GBP
APOSBIBA1021-1G APOSBIBA1021-5G APOSBIBA1021-10G
H-L-Dap(Boc)-OMe·HCl
H-L-Dap(boc)-OMe·HCl
Formula: C₉H₁₉ClN₂O₄
MW: 254.711 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00798684
CAS Number: 114559-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR