Fmoc-Lys(Me3)-OH chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Fmoc-Lys(Me)₃-OH chloride, Fmoc-Lys(Me)3-OH Chloride, Fmoc-Lys(Me)3-OH Chloride

BIBA1011-100MG BIBA1011-250MG
APOSBIBA1011-100MGEA 41 GBP
APOSBIBA1011-100MG APOSBIBA1011-250MG
Fmoc-Lys(Me3)-OH chloride
Fmoc-Lys(Me3)-OH chloride
Formula: C₂₄H₃₁ClN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00672333
CAS Number: 201004-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR