Bisacrylamide ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

BIB6418-100G BIB6418-1KG BIB6418-250G
APOSBIB6418-100GEA 51.5 GBP
APOSBIB6418-100G APOSBIB6418-1KG APOSBIB6418-250G
Bisacrylamide ≥98%
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR