Semicarbazide hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

BIB6269-1KG BIB6269-500G BIB6269-100G
APOSBIB6269-1KGEA 94.9 GBP
APOSBIB6269-1KG APOSBIB6269-500G APOSBIB6269-100G
Semicarbazide hydrochloride
Semicarbazide hydrochloride
Formula: CH₆ClN₃O
MW: 111.53 g/mol
Melting Pt: 175…177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013009
CAS Number: 563-41-7
ADR: 6.1 III
Merck Index: 12,08588

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR