5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

BIB6006-100MG BIB6006-1G BIB6006-500MG
APOSBIB6006-100MGEA 24.2 GBP
APOSBIB6006-100MG APOSBIB6006-1G APOSBIB6006-500MG
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP Red)
Formula: C₈H₆BrClNO₄P·C₇H₉N
Storage Temperature: Freezer
MDL Number: MFCD00210023
CAS Number: 6769-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR