Sodium chloride Pure BP, Ph. Eur., JP, USP, pharma grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Saline, NaCl, Sea salt, Halite

BIA1371-1KG BIA1371-5KG
APOSBIA1371-1KGEA 31.5 GBP
APOSBIA1371-1KG APOSBIA1371-5KG
Sodium chloride Pure BP, Ph. Eur., JP, USP, pharma grade
Sodium chloride
Formula: NaCl
Boiling Pt: 1461 °C
Melting Pt: 801 °C
Density: 2.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR