4-Aminobenzoic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1

BIA0601-100G BIA0601-25G
APOSBIA0601-100GEA 34.7 GBP
APOSBIA0601-100G APOSBIA0601-25G
4-Aminobenzoic acid 99%
4-Aminobenzoic acid

Intermediate for dyestuff, pigments, pharmaceuticals.

Formula: C₇H₇NO₂
Melting Pt: 186…189 °C
Density: 3.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007894
CAS Number: 150-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR