Chlortetracycline hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

BI7654-10G BI7654-100G BI7654-25G
APOSBI7654-10GEA 36.8 GBP
APOSBI7654-10G APOSBI7654-100G APOSBI7654-25G
Chlortetracycline hydrochloride
Chlortetracycline hydrochloride
Formula: C₂₂H₂₃ClN₂O₈·HCl
Melting Pt: 210 °C
Storage Temperature: Freezer
MDL Number: MFCD26142699
CAS Number: 64-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR