5-Bromo-2'-deoxyuridine (broxuridine)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

BI2048-1G BI2048-5G BI2048-25G
APOSBI2048-1GEA 17.9 GBP
APOSBI2048-1G APOSBI2048-5G APOSBI2048-25G
5-Bromo-2'-deoxyuridine (broxuridine)
5-Bromo-2'-deoxyuridine (Broxuridine)

Thymidine analogue used as a mutagen in genetic research.

Formula: C₉H₁₁BrN₂O₅
Melting Pt: 190…194 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR