Anti-KCNQ4 Mouse Monoclonal Antibody [clone: N43/6]

Supplier: ANTIBODIES.COM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A305021-100
ANTIA305021-100EA 375 GBP
ANTIA305021-100
Anti-KCNQ4 Mouse Monoclonal Antibody [clone: N43/6]
Antibodies
Mouse monoclonal [N43/6] antibody to KCNQ4 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: KCNQ4
Clonality: Monoclonal
Clone: N43/6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR