Anti-StAR Rabbit Recombinant Antibody [clone: STAR/3915R]

Supplier: ANTIBODIES.COM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A278067-100
ANTIA278067-100EA 355 GBP
ANTIA278067-100
Anti-StAR Rabbit Recombinant Antibody [clone: STAR/3915R]
Antibodies
Recombinant rabbit monoclonal [STAR/3915R] antibody to StAR for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: StAR
Clonality: Recombinant
Clone: STAR/3915R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR