Precision balances, advanced-level, Apollo series
Supplier: AnD Instruments Ltd (A&D)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GX-1603A GF-1003A GF-1202A GF-123A GF-1603A GF-2002A GF-203A GF-3002A GF-303A GF-403A GF-603A GF-10001A GF-6001A GF-10002A GF-4002A GX-1003A GX-10001A GX-10002A
ANDAGX-1603AEA 0 GBP
ANDAGX-1603A ANDAGF-1003A ANDAGF-1202A ANDAGF-123A ANDAGF-1603A ANDAGF-2002A ANDAGF-203A ANDAGF-3002A ANDAGF-303A ANDAGF-403A ANDAGF-603A ANDAGF-10001A ANDAGF-6001A ANDAGF-10002A ANDAGF-4002A ANDAGX-1003A ANDAGX-10001A ANDAGX-10002A
Precision balances, advanced-level, Apollo series
Balances and Scales
The A&D Apollo Precision Series of balances improve productivity and efficiency of your workflows with their user-friendly and innovative features, designed to provide reliable results across a range of environments and applications.
  • Impact shock detection (ISD) provides feedback to prevent damage to the weight sensor
  • Electronically controlled load (ECL) that enables precision management without using an external weight
  • Flow rate display (FRD) make calculating and recording flow rates easier and more accurate
  • Automatic self calibration in response to
More Product Information
Order Now
SPECIFICATIONS
Operating temperature range 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C 5...40 °C
Environmentally Preferable
Communication interface RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB RS232C and USB
Voltage 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC 30 VAC