Precision balances, advanced-level, Apollo series
Supplier: AnD Instruments Ltd (A&D)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GX-1603A GF-1003A GF-1202A GF-123A GF-1603A GF-2002A GF-203A GF-3002A GF-303A GF-403A GF-603A GF-10001A GF-6001A GF-10002A GF-4002A GX-1003A GX-10001A GX-10002A
ANDAGX-1603AEA 0 GBP
ANDAGX-1603A ANDAGF-1003A ANDAGF-1202A ANDAGF-123A ANDAGF-1603A ANDAGF-2002A ANDAGF-203A ANDAGF-3002A ANDAGF-303A ANDAGF-403A ANDAGF-603A ANDAGF-10001A ANDAGF-6001A ANDAGF-10002A ANDAGF-4002A ANDAGX-1003A ANDAGX-10001A ANDAGX-10002A
Precision balances, advanced-level, Apollo series
Balances and Scales
The A&D Apollo Precision Series of balances improve productivity and efficiency of your workflows with their user-friendly and innovative features, designed to provide reliable results across a range of environments and applications.
  • Impact shock detection (ISD) provides feedback to prevent damage to the weight sensor
  • Electronically controlled load (ECL) that enables precision management without using an external weight
  • Flow rate display (FRD) make calculating and recording flow rates easier and more accurate
  • Automatic self calibration in response to
More Product Information