Sodium hydrogen carbonate 75 g/l aqueous solution

Supplier: Alpha Diagnostics International
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium bicarbonate

BICA0002012
ALPDBICA0002012EA 0 GBP
ALPDBICA0002012
Sodium hydrogen carbonate 75 g/l aqueous solution
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR