2100 Bioanalyzer Instrument
Supplier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G2939BA
AGLSG2939BAEA 0 GBP
AGLSG2939BA
2100 Bioanalyzer Instrument
Electrophoresis Systems Capillary Electrophoresis Systems
Sizing, quantification, and sample quality control of DNA, RNA, and proteins on a single platform.
  • Replaceable electrode cartridges that can be easily exchanged for contamination-free switching between methods
  • The system features minimum exposure to hazardous materials to ensure operator safety
  • The 2100 Bioanalyzer instrument is compliant with European RoHS directives
  • For GLP/GMP environments, the 2100 Bioanalyzer system offers IQ and OQ services, as well as features to enable 21 CF Part 11 compliance
The 2100 Bioanalyzer system is an establishedMore Product Information