Cloned Pfu DNA polymerase alternative detergents
Supplier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
600355 600357
AGLS600355EA 318 GBP
AGLS600355 AGLS600357
Cloned Pfu DNA polymerase alternative detergents
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Cloned Pfu DNA Polymerase for high fidelity and difficult, GC-Rich PCR.
  • Highly thermostable Pfu DNA polymerase possessing 3' to 5' exonuclease proofreading activity that enables the polymerase to correct nucleotide-misincorporation errors (1 error per 770000 bp)
  • Reformulated buffer system for increased economy with the same high performance as the original Pfu DNA polymerase
  • Blunt ends
Using Cloned Pfu DNA polymerase in PCR experiments results in blunt-ended products, which are ideal for cloning into blunt-ended vectors. Cloned Pfu DNA Polymerase, derived from the hyperthermophilic archaea Pyrococcus furiosus exhibits superior thermostability and proofreading properties.
Caution: For research use only. Not for use in diagnostic procedures.