High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5067-5584 5067-5585 5067-5587 5067-5592 5067-5593 5067-5594 5067-5603
AGLS5067-5584EA 274 GBP
AGLS5067-5584 AGLS5067-5585 AGLS5067-5587 AGLS5067-5592 AGLS5067-5593 AGLS5067-5594 AGLS5067-5603
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Electrophoresis Systems Capillary Electrophoresis Systems
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
  • The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
  • Results are obtained quickly, in approximately 1 minute per sample
  • Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
  • Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • Reliable sample quality control is provided for next-
More Product Information