AdEasy Adenoviral vector systems
Supplier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
240005 240006 240007 240008 240009 240010 240081 240082 240085
AGLS240005EA 395 GBP
AGLS240005 AGLS240006 AGLS240007 AGLS240008 AGLS240009 AGLS240010 AGLS240081 AGLS240082 AGLS240085
AdEasy Adenoviral vector systems
Vectors, Plasmids and Libraries
The AdEasy system saves you a month of work over traditional methods by producing the recombinant adenoviral plasmid by homologous recombination in E. coli. The AdEasy XL system includes BJ5183 cells pre-transformed with the pAdEasy-1 plasmid, this feature dramatically decreases background caused by the non-recombinant shuttle plasmid.
  • 300% more recombinants with specialty competent cells
  • Adherent AD-293 cells improve viral packaging
  • Infects dividing and non-dividing cells
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.