Beakers, low form, stainless steel
Supplier: Agencinox
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2100B 2025 2050 2100 2200 2300 2500 2100A 2200A 2500A 2200AB 2100AB 2500B 2300AB 2200B 2300B 2010B 2025B 2050B 2010AB 2025AB 2050AB 2500AB 2010 2300A
AGEN2100BEA 190 GBP
AGEN2100B AGEN2025 AGEN2050 AGEN2100 AGEN2200 AGEN2300 AGEN2500 AGEN2100A AGEN2200A AGEN2500A AGEN2200AB AGEN2100AB AGEN2500B AGEN2300AB AGEN2200B AGEN2300B AGEN2010B AGEN2025B AGEN2050B AGEN2010AB AGEN2025AB AGEN2050AB AGEN2500AB AGEN2010 AGEN2300A
Beakers, low form, stainless steel
Beakers
Stainless steel.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.