Instrument trays, rectangular
Supplier: Agencinox
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1252 1242
AGEN1252EA 203 GBP
AGEN1252 AGEN1242
Instrument trays, rectangular
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Excellent thermal resistance