Abgene™ 96 Well Polypropylene Sample Processing and Storage Microplates
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AB-0796
ADVAAB-0796EA 0 GBP
ADVAAB-0796
Abgene™ 96 Well Polypropylene Sample Processing and Storage Microplates
Microplates Deep Well Microplates
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
  • Alphanumeric grid-referencing and industry standard footprint
  • Sealing options: Adhesive seals, heat seals and
More Product Information