Liquid viewer, dimmable, Apollo II
Supplier: ADELPHI MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6031515
ADLM6031515EA 1544.81 GBP
ADLM6031515
Liquid viewer, dimmable, Apollo II
Lights Light Boxes
Designed for use in pharmaceutical and laboratory environments, the Apollo II Dimmable Liquid Viewer provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
  • Light level between 1,200 and 3,500 Lux: adjustable by dimmer control, depending on the light in the room
  • Height and angle adjustable
  • Intuitive assembly; no tools required
The Apollo II Dimmable Liquid Viewer can quickly and easily be folded for storage, with the inspection surfaces folding together to reduce chance of damage. It is packaged in sustainablyMore Product Information