6-Bromoindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 6-Bromo-1H-indole

AS7108G25 AS7108G5
ACTIAS7108G25EA 0 GBP
ACTIAS7108G25 ACTIAS7108G5
6-Bromoindole
6-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR