2-Chloro-4-methylpyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS5149G5
ACTIAS5149G5EA 0 GBP
ACTIAS5149G5
2-Chloro-4-methylpyrimidine
2-Chloro-4-methylpyrimidine
Formula: C₅H₅ClN₂
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR