4-(Tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-(Tri-n-butylstannyl)pyridine

AS50170G1 AS50170G25 AS50170G5
ACTIAS50170G1EA 0 GBP
ACTIAS50170G1 ACTIAS50170G25 ACTIAS50170G5
4-(Tributylstannyl)pyridine
4-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
MDL Number: MFCD01319071
CAS Number: 124252-41-1
UN: 2788
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR