6-Indoleboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Indole-6-boronic acid

AS50065G1 AS50065G25 AS50065G5
ACTIAS50065G1EA 0 GBP
ACTIAS50065G1 ACTIAS50065G25 ACTIAS50065G5
6-Indoleboronic acid
6-Indoleboronic acid
Formula: C₈H₈BNO₂
MDL Number: MFCD03095176
CAS Number: 147621-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR