(4-Bromo-2-pyridinyl)methanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Bromo-2-(hydroxymethyl)pyridine, 4-Bromo-2-pyridinemethanol, (4-Bromopyridin-2-yl)methanol

AS31156G1 AS31156G5
ACTIAS31156G1EA 0 GBP
ACTIAS31156G1 ACTIAS31156G5
(4-Bromo-2-pyridinyl)methanol
(4-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
MW: 188.0200 g/mol
MDL Number: MFCD09800557
CAS Number: 131747-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR