2-Chloro-4-hydrazinopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS31013G1
ACTIAS31013G1EA 0 GBP
ACTIAS31013G1
2-Chloro-4-hydrazinopyridine
2-Chloro-4-hydrazinopyridine
Formula: C₅H₆ClN₃
MDL Number: MFCD08460247
CAS Number: 700811-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR