3,3,3-Trifluoro-1-propanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,3,3-Trifluoropropan-1-ol

AS24897G5
ACTIAS24897G5EA 0 GBP
ACTIAS24897G5
3,3,3-Trifluoro-1-propanol
3,3,3-Trifluoro-1-propanol
Formula: C₃H₅F₃O
Boiling Pt: 96…98 °C (1013 hPa)
Density: 1.3739 g/cm³ (25 °C)
MDL Number: MFCD00190649
CAS Number: 2240-88-2
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR