4-(Dimethylamino)cinnamic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-(Dimethylamino)cinnamic Acid

AS24625G100 AS24625G25 AS24625G5
ACTIAS24625G100EA 218 GBP
ACTIAS24625G100 ACTIAS24625G25 ACTIAS24625G5
4-(Dimethylamino)cinnamic acid
4-(Dimethylamino)cinnamic acid
Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
MDL Number: MFCD00004397
CAS Number: 1552-96-1
EINECS: 216-299-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR