2-(Bromomethyl)naphthalene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS24437G100 AS24437G25
ACTIAS24437G100EA 0 GBP
ACTIAS24437G100 ACTIAS24437G25
2-(Bromomethyl)naphthalene
2-(Bromomethyl)naphthalene
Formula: C₁₁H₉Br
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004123
CAS Number: 939-26-4
EINECS: 213-359-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR