4-Bromo-1,8-naphthalic anhydride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 6-Bromobenzo[de]isochromene-1,3-dione

AS24212G100 AS24212G25
ACTIAS24212G100EA 0 GBP
ACTIAS24212G100 ACTIAS24212G25
4-Bromo-1,8-naphthalic anhydride
4-Bromo-1,8-naphthalic anhydride
Formula: C₁₂H₅BrO₃
MW: 277.0703 g/mol
MDL Number: MFCD00006927
CAS Number: 81-86-7
EINECS: 201-382-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR