5-(3-Chlorophenyl)oxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS23816G5
ACTIAS23816G5EA 390 GBP
ACTIAS23816G5
5-(3-Chlorophenyl)oxazole
5-(3-Chlorophenyl)oxazole
Formula: C₉H₆ClNO
MW: 179.6000 g/mol
MDL Number: MFCD00084974
CAS Number: 89808-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR