Methyl 5-fluoro-2-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS23571G25 AS23571G5
ACTIAS23571G25EA 0 GBP
ACTIAS23571G25 ACTIAS23571G5
Methyl 5-fluoro-2-nitrobenzoate
Methyl 5-fluoro-2-nitrobenzoate
Formula: C₈H₆FNO₄
MDL Number: MFCD03425619
CAS Number: 393-85-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR