2,3,5-Trifluoro-4-methylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS23123G1 AS23123G5
ACTIAS23123G1EA 0 GBP
ACTIAS23123G1 ACTIAS23123G5
2,3,5-Trifluoro-4-methylbenzoic acid
2,3,5-Trifluoro-4-methylbenzoic acid
Formula: C₈H₅F₃O₂
MDL Number: MFCD00456800
CAS Number: 773869-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR