2-(2-Aminoethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine

AS23118G100 AS23118G25
ACTIAS23118G100EA 0 GBP
ACTIAS23118G100 ACTIAS23118G25
2-(2-Aminoethyl)pyridine
2-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
Boiling Pt: 49…50 °C (0.03 mmHg)
Density: 1,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR