3'-Fluoro-4'-hydroxyacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS23052G1 AS23052G100 AS23052G25 AS23052G5
ACTIAS23052G1EA 0 GBP
ACTIAS23052G1 ACTIAS23052G100 ACTIAS23052G25 ACTIAS23052G5
3'-Fluoro-4'-hydroxyacetophenone
3'-Fluoro-4'-hydroxyacetophenone
Formula: C₈H₇FO₂
MDL Number: MFCD00215840
CAS Number: 403-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR