2-Bromo-3-fluorophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS22843G1 AS22843G25 AS22843G5
ACTIAS22843G1EA 0 GBP
ACTIAS22843G1 ACTIAS22843G25 ACTIAS22843G5
2-Bromo-3-fluorophenol
2-Bromo-3-fluorophenol
Formula: C₆H₄BrFO
MDL Number: MFCD04037220
CAS Number: 443-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR