2-Chloro-4-fluoro-5-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

AS22720G1 AS22720G5
ACTIAS22720G1EA 0 GBP
ACTIAS22720G1 ACTIAS22720G5
2-Chloro-4-fluoro-5-nitroanisole
2-Chloro-4-fluoro-5-nitroanisole
Formula: C₇H₅ClFNO₃
MDL Number: MFCD03425810
CAS Number: 84478-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR