2,5-Dichloronicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS22712G1 AS22712G25 AS22712G5
ACTIAS22712G1EA 0 GBP
ACTIAS22712G1 ACTIAS22712G25 ACTIAS22712G5
2,5-Dichloronicotinic acid
2,5-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD01861979
CAS Number: 59782-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR