4-Fluoro-2-iodoaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS22394G100 AS22394G25
ACTIAS22394G100EA 0 GBP
ACTIAS22394G100 ACTIAS22394G25
4-Fluoro-2-iodoaniline
4-Fluoro-2-iodoaniline
Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR