2-Fluoro-4-(methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS22330G1 AS22330G25 AS22330G5
ACTIAS22330G1EA 0 GBP
ACTIAS22330G1 ACTIAS22330G25 ACTIAS22330G5
2-Fluoro-4-(methylsulphonyl)benzoic acid
2-Fluoro-4-(methylsulphonyl)benzoic acid
Formula: C₈H₇FO₄S
MDL Number: MFCD11615492
CAS Number: 142994-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR